Learn More
Sigma Aldrich Fine Chemicals Biosciences Thermal Conductivity Fib
Catalog No. 502909112
Encompass
SRM 1450e cert SRM 1450e SDSThis Standard Reference Material (SRM) is intended primarily for use in the measurement of the thermal conductivity or thermal resistance of insulation materials. SRM 1450e is a high-density fibrous glass board certified for bulk density and thermal conductivity. The SRM can be used in conjunction with ASTM C177 or ASTM C518. A unit of SRM 1450e consists of a square panel of fibrous glass and phenolic binder molded into a semi-rigid board. The nominal dimensions of a unit are 610 mm x 610 mm x 25 mm.
Catalog No. 50-290-9112 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. NIST1450E
SRM 1450e cert SRM 1450e SDSThis Standard Reference Material (SRM) is intended primarily for use in the measurement of the thermal conductivity or thermal resistance of insulation materials. SRM 1450e is a high-density fibrous glass board certified for bulk density and thermal conductivity. The SRM can be used in conjunction with ASTM C177 or ASTM C518. A unit of SRM 1450e consists of a square panel of fibrous glass and phenolic binder molded into a semi-rigid board. The nominal dimensions of a unit are 610 mm x 610 mm x 25 mm.