Learn More
TA Instruments Thermal Conductivity Kit for MDSC, Measures Thermal Conductivity of Insulating Materials, Range 0. 1–1. 5 W/°C · m, Covers Polymers, Ceramics, Glasses, Requires MDSC, Compatible with MDSC
Catalog No. 5000580608 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments Thermal Conductivity Kit for MDSC permits the measurement of thermal conductivity of insulating materials in the range from 0.1 to 1.5 W/°C·m, which generally covers polymers, ceramics, and glasses. This kit requires the use of MDSC to make the thermal conductivity measurement.
Catalog No. 50-005-80608 Supplier TA Instruments Supplier No. 915064.901
Waters TA Instruments Thermal Conductivity Kit for MDSC permits the measurement of thermal conductivity of insulating materials in the range from 0.1 to 1.5 W/°C·m, which generally covers polymers, ceramics, and glasses. This kit requires the use of MDSC to make the thermal conductivity measurement.