Learn More
AirSea USA ThermalControl Unit TC-100 (405mm x 395 x 575mm)
Catalog No. 501534817
Encompass_Preferred
This AirSea thermal control unit has a dry ice capacity of 75 kg/165 lbs. (Approx). Dry Ice Loss Rate: 481 Grams per hour. Thermal hold time: Up to 150 hours to 6.25 days. Loss Rates we calculated with a constant external ambient temperature of 30C. *Dry ice loss rates where shown are for guidance only. Results will vary in different environments.
Catalog No. 50-153-4817 Supplier AirSea USA Supplier No. 524
This AirSea thermal control unit has a dry ice capacity of 75 kg/165 lbs. (Approx). Dry Ice Loss Rate: 481 Grams per hour. Thermal hold time: Up to 150 hours to 6.25 days. Loss Rates we calculated with a constant external ambient temperature of 30C. *Dry ice loss rates where shown are for guidance only. Results will vary in different environments.