Learn More
Electron Microscopy Sciences DT Cryo-Tag Labels 3/8" dia, Orange, 0.5 mL tubes
SDP
Supplier:  Electron Microscopy Sciences 7757731
Encompass_Preferred
Direct thermal printer labela are temp resistant from -196 degree C to 70 degree C and freezable in liquid nitrogen and vapor phase nitrogen. Tough – They adhere to most plastics, glass and metals without cracking, peeling or degrading. Also resistant to scuffing and smearing. Tubes labeled with Direct Thermal CryoTags easily slide in and out of centrifuge rotors without binding. Labels accept printing from direct thermal printers such as Dymo LabelWriters.
Color: Orange
Use: 0.5 mL tubes
Catalog No. 50-192-7178