Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Thermal Fuse 250 V, 17 A

Catalog No. NC9456278 Shop All Thermo Scientific Products
Encompass_Preferred

Thermal Fuse 250 V, 17 A.

Catalog No. NC9456278 Supplier Thermo Scientific Supplier No. 285599
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thermal Fuse 250 V, 17 A.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.