Learn More
USA Scientific Inc Thermal-Lok 4-Position Dry Heat Bath
Catalog No. NC1828555
Encompass
Thermal-Lok four block position dry heat bath (without blocks), digital display, 5C above ambient to 150C, 1-999 minute timer
Catalog No. NC1828555 Supplier USA Scientific Inc Supplier No. 25101104
Thermal-Lok four block position dry heat bath (without blocks), digital display, 5C above ambient to 150C, 1-999 minute timer