Learn More
Electron Microscopy Sciences Thermal Microbeads 2.5-3 mm 1 LT
SDP
Catalog No. 501927578
Encompass_Preferred
Thermal Microbeads effectively heat and cool samples in most water and dry baths. Use of the microbeads reduces sample contamination in applications requiring water and ice and provides support for tubes and bottles, eliminating the need for floating racks and bottle weights. The autoclavable and solvent resistant microbeads can be used in water/dry baths, ice pans/buckets, microbead chambers, and microbead bags.
Catalog No. 50-192-7578 Supplier Electron Microscopy Sciences Supplier No. 6860510
Thermal Microbeads effectively heat and cool samples in most water and dry baths. Use of the microbeads reduces sample contamination in applications requiring water and ice and provides support for tubes and bottles, eliminating the need for floating racks and bottle weights. The autoclavable and solvent resistant microbeads can be used in water/dry baths, ice pans/buckets, microbead chambers, and microbead bags.