Learn More
Grainger Thermal Mittens, Zetex Plus, One Size Fits Most, 1000°F Max. Temp., 14" Length, Yellow, 1 PR
Catalog No. NC1744880
Encompass_Preferred
High Heat Protective Gloves, Mittens, and Covers and Multilayered Reversible Mittens and Mitten Covers. Mittens feature quilted Zetex Plus with wool lining. Mitten covers can be worn over top for increased protection and to extend the useful life of mittens. Both styles are reversible for extended wear life.
This item is not returnable. View return policy
Catalog No. NC1744880 Supplier Grainger Supplier No. 3GAE9
High Heat Protective Gloves, Mittens, and Covers and Multilayered Reversible Mittens and Mitten Covers. Mittens feature quilted Zetex Plus with wool lining. Mitten covers can be worn over top for increased protection and to extend the useful life of mittens. Both styles are reversible for extended wear life.