Learn More
Julabo USA Inc 10L Thermal P60 bath fluid -60…+250 °C in PRESTO only!
SDP
Catalog No. NC0276281
Encompass_Preferred
Bath fluid, Thermal P60, 10 L, For use in PRESTO highly dynamic temperature control systems only! Thermal P60 is a clear 5 cSt silicone fluid rated for use in JULABO PRESTO units only with a working temperature range of -60…+250 °C. This temperature range is only valid when used in PRESTO systems! The internal PRESTO cold oil overlay technology enables for safe operation above the fluid flashpoint.
Catalog No. NC0276281 Supplier Julabo USA Inc Supplier No. 8891412
Bath fluid, Thermal P60, 10 L, For use in PRESTO highly dynamic temperature control systems only! Thermal P60 is a clear 5 cSt silicone fluid rated for use in JULABO PRESTO units only with a working temperature range of -60…+250 °C. This temperature range is only valid when used in PRESTO systems! The internal PRESTO cold oil overlay technology enables for safe operation above the fluid flashpoint.