Learn More
Julabo USA Inc THERMAL P90 (10L) bath fluid, -90…+170 C; for use in PRESTO only
SDP
Catalog No. NC0355086
Encompass_Preferred
Temperature range -90...+170 deg.C. 2 cSt silicone based thermal fluid for use with Julabo Presto systems only.
Catalog No. NC0355086 Supplier Julabo USA Inc Supplier No. 8891414
Temperature range -90...+170 deg.C. 2 cSt silicone based thermal fluid for use with Julabo Presto systems only.