Learn More
Julabo USA Inc 5L Thermal P90 bath fluid -90…+170 °C in PRESTO only!
SDP
Catalog No. NC0430087
Encompass_Preferred
Bath fluid, Thermal P90, 5 L, For use in PRESTO highly dynamic temperature control systems only! Thermal P90 is a clear 2 cSt silicone fluid rated for use in JULABO PRESTO units only with a working temperature range of -90…+170 °C. This temperature range is only valid when used in PRESTO systems! The internal PRESTO cold oil overlay technology enables for safe operation above the fluid flashpoint.
Catalog No. NC0430087 Supplier Julabo USA Inc Supplier No. 8891415
Bath fluid, Thermal P90, 5 L, For use in PRESTO highly dynamic temperature control systems only! Thermal P90 is a clear 2 cSt silicone fluid rated for use in JULABO PRESTO units only with a working temperature range of -90…+170 °C. This temperature range is only valid when used in PRESTO systems! The internal PRESTO cold oil overlay technology enables for safe operation above the fluid flashpoint.