Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CoaguSense™ Thermal Paper

Catalog No. 23654010
Change view
Click to view available options
Quantity:
5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-654-010 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-654-010 Supplier CoaguSense™ Supplier No. PD99906
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CoaguSense™ Thermal Paper for use with PT2 Meter Portable Printer.

  • 5 rolls
  • For use with Coag-Sense™ Portable Printer

Specifications

Color Code White
Content And Storage Room Temperature
Type Paper
Includes 5 rolls of paper
Quantity 5
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.