Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arkray Thermal Printer Paper, 12 Rolls/Box
SDP

Catalog No. 21377295
Change view
Click to view available options
For Use With (Equipment):
Arkray Analyzer
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
21-377-295 Arkray Analyzer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Use only with compatible ARKRAY instruments. Store in a cool, dry place.
Catalog No. 21-377-295 Supplier Arkray Supplier No. OM98105
Add to Cart
Add to Cart
Use only with compatible ARKRAY instruments. Store in a cool, dry place.

ARKRAY Thermal Printer Paper produces sharp, accurate printouts for ARKRAY analyzers, ensuring clear records and dependable performance in laboratory settings.

  • Designed specifically for ARKRAY analyzers and thermal printers to ensure optimal compatibility.
  • Produces crisp, clear printouts for patient results, QC reports, and instrument logs.
  • Supports accurate documentation for regulatory compliance and record retention.
  • Durable print stability helps minimize fading over time for reliable result traceability.
  • Smooth surface coating promotes consistent image transfer and reduces printer wear.
  • Convenient roll format allows easy installation and continuous printing.
  • Ideal for high-volume or routine lab workflows requiring dependable output.
  • Engineered to minimize paper jams and printing interruptions.
  • Quality-tested to meet ARKRAY performance standards for clarity and reliability.
  • Helps laboratories maintain organized reporting, audit readiness, and smooth instrument operation.

Specifications

Adhesive Type Non-adhesive
Coating None
Color White
Finish Matte
Format Roll
Material Paper
Print Technology Direct Thermal, Thermal Transfer
Product Type Thermal Paper
Sterility Non-sterile
For Use With (Equipment) Arkray Analyzer
Length (English) 85 ft.
Width (English) 90 mm
Show More Show Less

Performance may vary depending on printer model and environmental conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.