Learn More
GA International Thermal-transfer, xylene and solvent-resistant resin ribbons 3.3" x 243' / 84mm x 74m Black 0.5" / 12.7mm
Catalog No. NC1372217
Encompass_Preferred
Xylene and solvent-resistant resin ribbons for thermal-transfer printers. When used with the appropriate label, the ribbon will provide a xylene and chemical-resistant printout. Range of sizes available, ensuring compatibility with most common barcode printer brands.
Catalog No. NC1372217 Supplier GA International Supplier No. XAR84X74C0.51JZ4
Xylene and solvent-resistant resin ribbons for thermal-transfer printers. When used with the appropriate label, the ribbon will provide a xylene and chemical-resistant printout. Range of sizes available, ensuring compatibility with most common barcode printer brands.