Learn More
Andwin Scientific THERMAL SEAL ADHESIVE FILM STL
SDP
Catalog No. NC9372417
Encompass_Preferred
The Thermaseal 50um-thick sealing film is made to enhance protection and leakage of either raised-rim PCR plates or standards plated during thermal cycling. This product has 50um clear polypropylene with a thicker 50um layer of stronger adhesive, two end tabs for easy positioning, and one sealing paddle for ensuring a firm seal. This product is free of DNase, RNase, and nucleic acid.
Catalog No. NC9372417 Supplier Andwin Scientific Supplier No. 05612520
The Thermaseal 50um-thick sealing film is made to enhance protection and leakage of either raised-rim PCR plates or standards plated during thermal cycling. This product has 50um clear polypropylene with a thicker 50um layer of stronger adhesive, two end tabs for easy positioning, and one sealing paddle for ensuring a firm seal. This product is free of DNase, RNase, and nucleic acid.