Learn More
Research Products International Corp Thermal Seal MiniStrips, Non-Sterile, 200 per Package
SDP
Catalog No. 501978143
Encompass
ThermalSeal MiniStrips provide a quick and cost effective method for selectively sealing one or two 8-well rows when using PCR plates for small runs. Made of resistant polypropylene film with a strong adhesive for firm bonding to plates.
Adhesive films are specially designed for use as microplate covers. Prevent sample evaporation and contamination in sensitive PCR applications. RNase/DNase free. It is thermostable and functional from -40°C to +120°C at 75% humidity.
Strips measure 19.0 x 103.2 mm, including two removable positioning tabs. Length without tabs is 82.6 mm.
ThermalSeal is a Trademark of Excel Scientific, Inc.
Catalog No. 50-197-8143 Supplier Research Products International Corp Supplier No. 202509
ThermalSeal MiniStrips provide a quick and cost effective method for selectively sealing one or two 8-well rows when using PCR plates for small runs. Made of resistant polypropylene film with a strong adhesive for firm bonding to plates.
Adhesive films are specially designed for use as microplate covers. Prevent sample evaporation and contamination in sensitive PCR applications. RNase/DNase free. It is thermostable and functional from -40°C to +120°C at 75% humidity.
Strips measure 19.0 x 103.2 mm, including two removable positioning tabs. Length without tabs is 82.6 mm.
ThermalSeal is a Trademark of Excel Scientific, Inc.