Learn More
Cole-Parmer Thermal Transfer Compound, 4 oz Tube
Supplier:  Cole-Parmer 9378561
Encompass_Preferred
Silicone-free heat sink compound increases thermal conductivity in thermo wells for faster response time in comparison to air. This heat sink grease is also designed to transfer heat away from heat-generating components.
Catalog No. NC9081297