Learn More
PDC Thermal Transfer label, Paper, Permanent, 3800/Roll, 6 Rolls/Box
Catalog No. 21235883
Change view
Click to view available options
Quantity:
3800/Roll, 6 Rolls/Box
Catalog No. Quantity
21-235-883 3800/Roll, 6 Rolls/Box
1 options
Catalog No. 21-235-883 Supplier PDC Supplier No. TTR114212
Add to Cart
Add to Cart
Reliable paper thermal transfer labels with permanent adhesive, designed for dependable use in healthcare and laboratory labeling.
  • Engineered by PDC for consistent, high-quality printing in healthcare and laboratory settings
  • Thermal transfer paper construction supports sharp, scannable barcodes and clear text
  • White face stock enhances readability and professional labeling standards
  • Permanent adhesive provides secure bonding to tubes, vials, and containers
  • 3 in. core design ensures compatibility with standard thermal transfer printers
  • Each roll delivers 3,800 labels and sold as 6 rolls per box to meet high-volume needs
Specifications
Adhesive Type Permanent
Coating None
Color White
Finish Matte
Format Label
Material Paper
Printable Sides Front
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
Quantity 3800/Roll, 6 Rolls/Box
Dimensions (L x W) 2 1/2 x 1 1/4 in.
Length (English) 1.25 in.
Width (English) 2.5 in.
Width (Metric) 63.5 mm