Learn More
Brady™ Thermal Transfer Printable Labels, B-430
Used to identify property from components to finished products. Brady™ Thermal Transfer Printable Labels are made of clear polyester.
Supplier:  Brady™ THT3943010
 View more versions of this product
Catalog No. 19-109-329
Add to Cart
Description
Description
  • For use with thermal transfer printers
  • Made of BradyBondz™ translucent, high adhesion polyester material
Specifications
Specifications
Autoclavable
Recommended ribbon series: R4400; R4900; R6000; R6200
None
Gloss
Polyester (B-430)
Thermal Transfer
Non-sterile
BBP81, i5100, i7100, PR Plus, IP Series, THT 76 mm Core Printers, i7500
3
THT-39-430-10
10,000/Pk.
25.40 mm (W) x 12.70 mm (H)
-94°F to +212°F
0.003 in.
15.88 mm
0.5 in.
25.4 mm
0.5 in.
THT-39-430-10
UL Recognized; CSA Accepted
Acrylic, Permanent
Clear
Roll
Single-side Printable
Thermal Transfer Printer Label
Asset Tracking, Barcode Labeling, Component and Equipment Identification, General and Industrial Identification, Rating Plates
27.31 mm
-70°C
10000 Label(s) / Roll
Rectangle
25.4 x 12.7 mm (W x H)
-70°C to +100°C
0.076 mm
12.70 x 25.40 mm
1 in.
25.40 mm
12.7 mm
Product Suggestions
Product Suggestions