Learn More
Sigma Aldrich Fine Chemicals Biosciences Thermal Transfer TNCryo-T
Catalog No. 502900549
Encompass
Works with most thermal transfer printersTemperature resistant from -196C to 80C liquid/vapor phase nitrogenPrint popular barcode formats for fast and easy sample identificationSolvent-resistant adhesive provides excellent cold temperature performanceResists degradation from scuffing chemicals moisture and temperature fluctuations
Catalog No. 50-290-0549 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7434582000EA
Works with most thermal transfer printersTemperature resistant from -196C to 80C liquid/vapor phase nitrogenPrint popular barcode formats for fast and easy sample identificationSolvent-resistant adhesive provides excellent cold temperature performanceResists degradation from scuffing chemicals moisture and temperature fluctuations