Learn More
GA International Thermal-Transfer Xylene & Solvent-Resistant Ribbon 2.5" x 243' / 64mm x 74m Black 0.5" / 12.7mm (plastic)
Catalog No. NC1498896
Encompass_Preferred
These resin-based ink ribbons for thermal-transfer printers will produce a solvent and xylene-resistant printout on labels used for histology and other applications that involve exposure to harsh chemicals and solvents. Available in a range of sizes, they are compatible with most common thermal-transfer printer brands and models*.
Catalog No. NC1498896 Supplier GA International Supplier No. XAR64X74CP0.51JZ4
These resin-based ink ribbons for thermal-transfer printers will produce a solvent and xylene-resistant printout on labels used for histology and other applications that involve exposure to harsh chemicals and solvents. Available in a range of sizes, they are compatible with most common thermal-transfer printer brands and models*.