Learn More
GA International Paper Labels for Thermal-Transfer Printers 0.9375" x 0.9375" / 23.8mm x 23.8mm Rectangle Green Apple From -20°C to +120°C (-4°F to +248°F)
Catalog No. NC2078908
Encompass_Preferred
General use paper labels for thermal-transfer printers. Provided with a permanent adhesive in a range of colors, they are ideal for use as shipping labels, color coding, price tagging, warehouses, logistics, and sorting facilities as well as general office and laboratory organization.
Catalog No. NC2078908 Supplier GA International Supplier No. GPB125C14GA
General use paper labels for thermal-transfer printers. Provided with a permanent adhesive in a range of colors, they are ideal for use as shipping labels, color coding, price tagging, warehouses, logistics, and sorting facilities as well as general office and laboratory organization.