Learn More
Braintree Scientific Thermalert Monitoring Thermometer
SDP
Supplier:  Braintree Scientific TH5
Encompass
Thermalert Monitoring Thermometer -. Long battery life - 1200 hours. This economical, easy to use thermometer is intended for continuous monitoring in the laboratory. The digital display reads to .1C in clinical range 25-45C. For safety, the unit is battery operated. Batteries last for 1200 hours. High stability - no calibration necessary. A variety of fast reading microprobes and special animal probes are available for the TH-5. TH-5 can be supplied with analog output for lab applications where strip chart recording is needed. Temperature range: 25 to 45C, Resolution: .1C, Instrument accuracy: .1C 1 digit, Calibration: Conforms to National Bureau of Standards tables Monograph 125, Readout: .5" liquid crystal, Batteries: 4 "C" cells, Battery life: 1200 hours continuous, Analog output: Available at extra cost. Approx. lOmV per C. **Sold for research purposes only and not for human use**
Catalog No. 50-195-6047