Learn More
USA Scientific Inc Thermal-Lok dry heat bath
Catalog No. 502745006
Encompass
Thermal-Lok dry heat bath, one block position (without blocks), digital display, 5C above ambient to 150C, 1-999 minute timer
Catalog No. 50-274-5006 Supplier USA Scientific Inc Supplier No. 25101101
Thermal-Lok dry heat bath, one block position (without blocks), digital display, 5C above ambient to 150C, 1-999 minute timer