Learn More
Andwin Scientific ThermalSeal 2 Film
SDP
Catalog No. NC1664384
Encompass_Preferred
These improved films for classic thermal cycling applications consist of a 50 µm polyester film with a 41 µm layer of acrylic adhesive. Compared to the original ThermalSeal films (below), ThermalSeal 2 films offer better adhesion, reduced evaporation, longer end tabs for easier handling, and less tendency to tear during removal from the plate. Each sealing film measures 79.4 x 139.7 mm and offers sufficient sealing area for all PCR plates. Length between the perforations with end tabs removed is 125.1 mm. (Not recommended for raised-rim plates; see ThermalSeal A films below.)
Catalog No. NC1664384 Supplier Andwin Scientific Supplier No. TS2100
These improved films for classic thermal cycling applications consist of a 50 µm polyester film with a 41 µm layer of acrylic adhesive. Compared to the original ThermalSeal films (below), ThermalSeal 2 films offer better adhesion, reduced evaporation, longer end tabs for easier handling, and less tendency to tear during removal from the plate. Each sealing film measures 79.4 x 139.7 mm and offers sufficient sealing area for all PCR plates. Length between the perforations with end tabs removed is 125.1 mm. (Not recommended for raised-rim plates; see ThermalSeal A films below.)