Learn More
Sigma Aldrich Fine Chemicals Biosciences ThermalSeal 2R sealing fil
Catalog No. 502900557
Encompass
These improved films for classic thermal cycling applications consist of a 50 mum polyester film with a 41 mum layer of acrylic adhesive. Compared to the original ThermalSeal(R) films ThermalSeal(R) 2 films offer better adhesion reduced evaporation longer end tabs for easier handling and less tendency to tear during removal from the plate. Each sealing film measures 79.4 x 139.7 mm and offers sufficient sealing area for all PCR plates. Length between the perforations with end tabs removed is 125.1 mm. ThermalSeal 2 sealing films are not pierceable.
Catalog No. 50-290-0557 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z768944100EA
These improved films for classic thermal cycling applications consist of a 50 mum polyester film with a 41 mum layer of acrylic adhesive. Compared to the original ThermalSeal(R) films ThermalSeal(R) 2 films offer better adhesion reduced evaporation longer end tabs for easier handling and less tendency to tear during removal from the plate. Each sealing film measures 79.4 x 139.7 mm and offers sufficient sealing area for all PCR plates. Length between the perforations with end tabs removed is 125.1 mm. ThermalSeal 2 sealing films are not pierceable.