Learn More
Research Products International Corp ThermalSeal A Sealing Film, Non-Sterile, 100 per Package
SDP
Catalog No. 50550231
Encompass
ThermalSeal A Sealing Film features a thick layer of a specially formulated adhesive for improved protection from evaporation and leakage. Ideal for use with PCR plates and standard assay plates. Designed to fit inside the edge of raised rim plates to provide optimum sealing. Temperature range is -40°C to 120°C. Supplied with a plastic applicator. ThermalSeal A is a Trademark of Excel Scientific, Inc.
Advanced sealing
Thicker adhesive layer
DNase and RNase free
Catalog No. 50-550-231 Supplier Research Products International Corp Supplier No. 202545
ThermalSeal A Sealing Film features a thick layer of a specially formulated adhesive for improved protection from evaporation and leakage. Ideal for use with PCR plates and standard assay plates. Designed to fit inside the edge of raised rim plates to provide optimum sealing. Temperature range is -40°C to 120°C. Supplied with a plastic applicator. ThermalSeal A is a Trademark of Excel Scientific, Inc.
Advanced sealing
Thicker adhesive layer
DNase and RNase free