Learn More
Sigma Aldrich Fine Chemicals Biosciences ThermalSeal ATM sealing fi
Catalog No. 502901004
Encompass
ThermalSeal A sealing films are an advanced version of Excel Scientifics classic ThermalSeal A sealing films for PCR. ThermalSeal A films consist of 50 mum thick clear polypropylene with a thicker 50 mum layer of a stronger adhesive formulated for improved protection against evaporation and leakage during thermal cycling. Intended for non-real-time PCR applications ThermalSeal A films are designed to fit inside the edge of raised-rim PCR plates and provide more secure sealing of all wells because the center of the film does not extend over the plate rim. Two end tabs allow for easy positioning of the film on the plate but are perforated for easy removal if necessary.Each film measures 77.8 x 135.5 mm overall. Length between the perforations with end tabs removed is 118.1 mm. Each package of ThermalSeal A films includes one sealing paddle for ensuring a firm seal also compatible with raised-rim plates.Recommended for non-real-time PCR thermocycling applicationsStronger and t
Catalog No. 50-290-1004 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z723304100EA
ThermalSeal A sealing films are an advanced version of Excel Scientifics classic ThermalSeal A sealing films for PCR. ThermalSeal A films consist of 50 mum thick clear polypropylene with a thicker 50 mum layer of a stronger adhesive formulated for improved protection against evaporation and leakage during thermal cycling. Intended for non-real-time PCR applications ThermalSeal A films are designed to fit inside the edge of raised-rim PCR plates and provide more secure sealing of all wells because the center of the film does not extend over the plate rim. Two end tabs allow for easy positioning of the film on the plate but are perforated for easy removal if necessary.Each film measures 77.8 x 135.5 mm overall. Length between the perforations with end tabs removed is 118.1 mm. Each package of ThermalSeal A films includes one sealing paddle for ensuring a firm seal also compatible with raised-rim plates.Recommended for non-real-time PCR thermocycling applicationsStronger and t