Learn More
Research Products International Corp Plate Sealing Films, ThermalSeal, Clear, Sterile, 100 per Package
SDP
Catalog No. 50212461
Encompass
Adhesive films are specially designed for use as microplate covers. Prevent sample evaporation and contamination in sensitive PCR applications. ThermalSeal is polypropylene based and RNase/DNase free. It is thermostable and functional from -40°C to +120°C at 75% humidity. Supplied with a split backing for accurate plate positioning and secure sealing. ThermalSeal is a Trademark of Excel Scientific, Inc.
Prevent evaporation
Thermostable adhesive
For PCR
RNase and DNase free
Chemically resistant
Transparent
Quickly seals microplates
Catalog No. 50-212-461 Supplier Research Products International Corp Supplier No. 202501
Adhesive films are specially designed for use as microplate covers. Prevent sample evaporation and contamination in sensitive PCR applications. ThermalSeal is polypropylene based and RNase/DNase free. It is thermostable and functional from -40°C to +120°C at 75% humidity. Supplied with a split backing for accurate plate positioning and secure sealing. ThermalSeal is a Trademark of Excel Scientific, Inc.
Prevent evaporation
Thermostable adhesive
For PCR
RNase and DNase free
Chemically resistant
Transparent
Quickly seals microplates