Learn More
Bellco Glass, Inc. ThermalSeal Sterile,2mil For Microplates
SDP
Catalog No. 501945603
Encompass_Preferred
Sealing films prevent evaporation and protect samples during processing, storage, and shipment. They have superior quality film and uniform adhesive. With 2 perforated end tabs for accurate positioning. Removable end-tab backing for sealing around ends of plate. Films are packaged in zipper bags. Manufactured from 2mil polyester. 80x135mm, -40C to 120C, PCR, Co2 exchange-No, Pierceable-No, DN/RNase free-Yes
Catalog No. 50-194-5603 Supplier Bellco Glass, Inc. Supplier No. 440000600
Sealing films prevent evaporation and protect samples during processing, storage, and shipment. They have superior quality film and uniform adhesive. With 2 perforated end tabs for accurate positioning. Removable end-tab backing for sealing around ends of plate. Films are packaged in zipper bags. Manufactured from 2mil polyester. 80x135mm, -40C to 120C, PCR, Co2 exchange-No, Pierceable-No, DN/RNase free-Yes