Learn More
Bellco Glass, Inc. ThermalSeal RT, 2mil For Microplates
SDP
Catalog No. 501945610
Encompass_Preferred
Sealing films prevent evaporation and protect samples during processing, storage, and shipment. They have superior quality film and uniform adhesive. With 2 perforated end tabs for accurate positioning. Removable end-tab backing for sealing around ends of plate. Films are packaged in zipper bags. Manufactured from 2mil polyester. 83x143mm, -40C to 120C, real time PCR, Co2 exchange-No, Pierceable-Yes, DN/RNase free-Yes
Catalog No. 50-194-5610 Supplier Bellco Glass, Inc. Supplier No. 440001300
Sealing films prevent evaporation and protect samples during processing, storage, and shipment. They have superior quality film and uniform adhesive. With 2 perforated end tabs for accurate positioning. Removable end-tab backing for sealing around ends of plate. Films are packaged in zipper bags. Manufactured from 2mil polyester. 83x143mm, -40C to 120C, real time PCR, Co2 exchange-No, Pierceable-Yes, DN/RNase free-Yes