Learn More
Andwin Scientific ThermalSeal RT2 Plate Sealing Film, 100/cs
SDP
Catalog No. NC1664391
Encompass_Preferred
ThermalSeal RT sealing films combine an optically transparent polyester sealing film with a strong, ultra-smooth, non-absorbing, non-fluorescing medical-grade adhesive for superior performance in real-time PCR applications.
Catalog No. NC1664391 Supplier Andwin Scientific Supplier No. TSRT2100
ThermalSeal RT sealing films combine an optically transparent polyester sealing film with a strong, ultra-smooth, non-absorbing, non-fluorescing medical-grade adhesive for superior performance in real-time PCR applications.