Learn More
Sigma Aldrich Fine Chemicals Biosciences ThermalSeal RT2RRTM film n
Catalog No. 502900988
Encompass
ThermalSeal RT2RR(TM) 50 mum polyester sealing films for real-time PCR are sized to fit within the edges of raised-rim 96 well plates. The same consistent ultra-high optical clarity as Excels ThermalSeal RT(TM) sealing films makes possible more reproducible reliable and consistent DNA amplification measurements. An inert strong temperature-resistant adhesive assures reliable sealing around each well. Two end tabs assist in positioning the sealing film. Easy removal of the end tabs at perforated boundaries prevents lifting and higher evaporation rates that can occur with sealing films that overlap the plate rim. Recommended for specific DNA sequence detection in clinical diagnostics forensic science and basic research.Dimensions 77.8 by 130.8 mm. With end tabs removed length is 118.1 mm with 45 corners.Ultra-high optical clarityfit within plate rim to prevent evaporation due to sealing film liftingHeat resistant recommended for temperatures from-40 C to 120 C.
Catalog No. 50-290-0988 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z722553100EA
ThermalSeal RT2RR(TM) 50 mum polyester sealing films for real-time PCR are sized to fit within the edges of raised-rim 96 well plates. The same consistent ultra-high optical clarity as Excels ThermalSeal RT(TM) sealing films makes possible more reproducible reliable and consistent DNA amplification measurements. An inert strong temperature-resistant adhesive assures reliable sealing around each well. Two end tabs assist in positioning the sealing film. Easy removal of the end tabs at perforated boundaries prevents lifting and higher evaporation rates that can occur with sealing films that overlap the plate rim. Recommended for specific DNA sequence detection in clinical diagnostics forensic science and basic research.Dimensions 77.8 by 130.8 mm. With end tabs removed length is 118.1 mm with 45 corners.Ultra-high optical clarityfit within plate rim to prevent evaporation due to sealing film liftingHeat resistant recommended for temperatures from-40 C to 120 C.