Learn More
Andwin Scientific ThermalSeal RTS Optically Clear Silicone Adhesive Plate Sealing Film, 100/cs
SDP
Catalog No. NC1664309
Encompass_Preferred
ThermalSeal RTS™ sealing films are based on 50 µm polyolefin films with 50 µm inert encapsulated silicone adhesive. Sized to fit within the edges of raised-rim 96-well plates.
Catalog No. NC1664309 Supplier Andwin Scientific Supplier No. TSSRTQ100
ThermalSeal RTS™ sealing films are based on 50 µm polyolefin films with 50 µm inert encapsulated silicone adhesive. Sized to fit within the edges of raised-rim 96-well plates.