Learn More
Sigma Aldrich Fine Chemicals Biosciences ThermalSeal RTSTM Sealing
Catalog No. 502900558
Encompass
ThermalSeal RTS(TM) sealing film is a 50 mum thick polyolefin with 50 mum inert encapsulated silicone adhesive. They are optically clear with low autofluorescence and are especially suited for real-time qPCR storage and protein crystallization applications. The encapsulated silicone adhesive is non-tacky until pressed against the sealing surface at which time adhesive is released only in sealing areas to form the strongest available heat-resistant seal around each well on the plate. Adhesive on non-sealing areas of the film such as directly over sample wells remains encapsulated and inert.ThermalSeal RTS(TM) films are sized to fit within the edges of raised-rim 96-well plates. Their consistent high optical clarity makes possible reproducible reliable and consistent DNA amplification measurements and crystal detection. Two end tabs assist in positioning the film on the plate and the non-tacky adhesive surface simplifies handling. Easy removal of the end tabs at perforated boun
Catalog No. 50-290-0558 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z74225650EA
Add to Cart
Add to Cart
ThermalSeal RTS(TM) sealing film is a 50 mum thick polyolefin with 50 mum inert encapsulated silicone adhesive. They are optically clear with low autofluorescence and are especially suited for real-time qPCR storage and protein crystallization applications. The encapsulated silicone adhesive is non-tacky until pressed against the sealing surface at which time adhesive is released only in sealing areas to form the strongest available heat-resistant seal around each well on the plate. Adhesive on non-sealing areas of the film such as directly over sample wells remains encapsulated and inert.ThermalSeal RTS(TM) films are sized to fit within the edges of raised-rim 96-well plates. Their consistent high optical clarity makes possible reproducible reliable and consistent DNA amplification measurements and crystal detection. Two end tabs assist in positioning the film on the plate and the non-tacky adhesive surface simplifies handling. Easy removal of the end tabs at perforated boun