Learn More
Bellco Glass, Inc. AlumaSeal II N/Sterile,1.5mil For Microplates
SDP
Catalog No. 501945604
Encompass_Preferred
Sealing films prevent evaporation and protect samples during processing, storage, and shipment. They have superior quality film and uniform adhesive. With 2 perforated end tabs for accurate positioning. Removable end-tab backing for sealing around ends of plate. Films are packaged in zipper bags. Manufactured from 2mil polyester. 83x143mm, -80C to 120C, thermal cycling, Co2 exchange-No, Pierceable-Yes, DN/RNase free-Yes
Catalog No. 50-194-5604 Supplier Bellco Glass, Inc. Supplier No. 440000700
Add to Cart
Add to Cart
Sealing films prevent evaporation and protect samples during processing, storage, and shipment. They have superior quality film and uniform adhesive. With 2 perforated end tabs for accurate positioning. Removable end-tab backing for sealing around ends of plate. Films are packaged in zipper bags. Manufactured from 2mil polyester. 83x143mm, -80C to 120C, thermal cycling, Co2 exchange-No, Pierceable-Yes, DN/RNase free-Yes