Learn More
Sigma Aldrich Fine Chemicals Biosciences ThermalSealR films classic
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z369683100EA
Encompass
A 50 mum heat-resistant polypropylene sealing film with a 25 mum layer of acrylic adhesive designed for thermal cycling applications. Each sealing film measures 79.4 x 137.8 mm and offers sufficient sealing area for all PCR plates. Length between the perforations with end tabs removed is 123.1 mm. ThermalSeal sealing films are not pierceable. for applications where piercing with pipet tips or robotic probes is required for product recovery use AlumaSeal(R) sealing films. for real-time PCR applications where maximum optical clarity is required use ThermalSeal RT sealing films.features and Benefitsheat resistant recommended for temperatures from -40 C to 120 Ceasier to apply than aluminum foils no tendence to roll backcertified DNase- RNase- and nucleic-acid-free
Catalog No. 50-290-1198