Learn More
Corning™ ThermalTray™ Thermo-conductive Platforms
BIOPHARMA
Catalog No. 07210073 Shop All Corning Products
Change view
Click to view available options
For Use With (Equipment):
9L ice pan w/ liquid nitrogen
9L ice pan with crushed ice
Water bath
Type:
HP, High Profile
LP, Low Profile
SLP, Slim, Low Profile
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Type
07-210-073 9L ice pan w/ liquid nitrogen SLP, Slim, Low Profile
07-210-074 9L ice pan with crushed ice LP, Low Profile
07-210-075 Water bath HP, High Profile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 07-210-073 Supplier Corning™ Supplier No. 432073
This item is not returnable. View return policy
Support Corning CoolRack™ and CoolSink™ sample modules in liquid temperature sources such as melting ice, water baths, and liquid nitrogen.
Corning ThermalTray™ thermo-conductive platforms support CoolRack™ and CoolSink™ sample modules in liquid temperature sources such as melting ice, water baths and liquid nitrogen. Made of the same highly conductive alloy as CoolRack and CoolSink modules, ThermalTray platforms conduct the source temperature to the CoolRack or CoolSink and, ultimately, to your samples.
  • Stable, sturdy design makes them ideal for processing temperature-sensitive samples in melted ice baths or liquid nitrogen.
  • All modules may be autoclaved, high heat sterilized or decontaminated with bleach, alcohol or other disinfectants or lab detergents.
TRUSTED_SUSTAINABILITY
Specifications
Description ThermalTray Thermo-conductive platform
For Use With (Equipment) 9L ice pan w/ liquid nitrogen
Type SLP, Slim, Low Profile