Learn More
Andwin Scientific THERMANOX COVER SLIP 500/PK
SDP
Catalog No. 50949693
Encompass_Preferred
Specimen Preparation and Embedding Supplies: Wells, Optical Bottom Plates, Cell Culture Inserts: Cover Slips; Thermanox; TMX Coverslips: 22mm Round Thermanox, 500/bx
Catalog No. 50-949-693 Supplier Andwin Scientific Supplier No. 72283
Specimen Preparation and Embedding Supplies: Wells, Optical Bottom Plates, Cell Culture Inserts: Cover Slips; Thermanox; TMX Coverslips: 22mm Round Thermanox, 500/bx