Learn More
Alkali Scientific ThermaSeal™, Optically Clear Sealing Film, 100/pk
SDP
Catalog No. 502388983
Encompass_Preferred
ThermaSeal RTS™ Optically Clear, Silicone Adhesive provides an extremely strong seal to minimize evaporation. The silicone adhesive is non-tacky until pressure is applied to the sealing surface which then creates a secure bond around the wells and perimeter of the plate. Also ideal for dry samples (e.g. soil, seeds, insects, etc.) because the non-activated adhesive above the wells remains non-tacky. A roller or sealing paddle is highly recommended for use with this product to ensure consistent and adequate pressure is applied during the sealing process. Certified DNase- & RNase- free.
Catalog No. 50-238-8983 Supplier Alkali Scientific Supplier No. SF28ML100
ThermaSeal RTS™ Optically Clear, Silicone Adhesive provides an extremely strong seal to minimize evaporation. The silicone adhesive is non-tacky until pressure is applied to the sealing surface which then creates a secure bond around the wells and perimeter of the plate. Also ideal for dry samples (e.g. soil, seeds, insects, etc.) because the non-activated adhesive above the wells remains non-tacky. A roller or sealing paddle is highly recommended for use with this product to ensure consistent and adequate pressure is applied during the sealing process. Certified DNase- & RNase- free.