Learn More
Alkali Scientific ThermaSeal PCR MiniStrips™ Sealing Films, 4 Strips per Sheet, 2X8 Rows per Strip
SDP
Catalog No. 502388858
Encompass_Preferred
ThermalSeal MiniStrips sealing films are identical to ThermalSeal sealing films, but designed for sealing only one or two 8-well rows at a time on PCR plates or strip tubes. Two tabs on each strip for easy application and removal. Perforated tabs can be removed or sealed around plate edge. Heated resistant, recommended for temperatures from -40°C to 120°C. Certified DNase-, RNase-, and nucleic-acid-free.
Catalog No. 50-238-8858 Supplier Alkali Scientific Supplier No. SF30ML
ThermalSeal MiniStrips sealing films are identical to ThermalSeal sealing films, but designed for sealing only one or two 8-well rows at a time on PCR plates or strip tubes. Two tabs on each strip for easy application and removal. Perforated tabs can be removed or sealed around plate edge. Heated resistant, recommended for temperatures from -40°C to 120°C. Certified DNase-, RNase-, and nucleic-acid-free.