Learn More
Research Products International Corp ThermaSeal RTS Sealing Film for RT qPCR, Non-Sterile, 100/pk
SDP
Catalog No. 501977496
Encompass
ThermalSeal RTS Sealing Film has a unique layer of adhesive that remains non-tacky until pressed against a sealing surface. Adhesive layer is composed of 50um thick inert encapsulated silicone. Film creates optically clear protection barrier over sample area as heat resistant seal secures each well. Silicone adhesive forms the strongest available seal for evaporation prevention. Non-tacking adhesive simplifies handling prior to sealing. ThermalSeal RTS film is sized to fit within the edges of raised rim 96-well plates and is compatible with standard plates. Temperature resistant from -80° C to +105° C. ThermalSeal RTS is a Trademark of Excel Scientific, Inc.
Excellent chemical resistance
Extra Strong, Non-tacky Adhesive
Suitable for storage of dry samples
Catalog No. 50-197-7496 Supplier Research Products International Corp Supplier No. 202547
ThermalSeal RTS Sealing Film has a unique layer of adhesive that remains non-tacky until pressed against a sealing surface. Adhesive layer is composed of 50um thick inert encapsulated silicone. Film creates optically clear protection barrier over sample area as heat resistant seal secures each well. Silicone adhesive forms the strongest available seal for evaporation prevention. Non-tacking adhesive simplifies handling prior to sealing. ThermalSeal RTS film is sized to fit within the edges of raised rim 96-well plates and is compatible with standard plates. Temperature resistant from -80° C to +105° C. ThermalSeal RTS is a Trademark of Excel Scientific, Inc.
Excellent chemical resistance
Extra Strong, Non-tacky Adhesive
Suitable for storage of dry samples