Learn More
Andwin Scientific THERMASEAL SEALING FILM 100/PK
SDP
Catalog No. NC9333898
Encompass_Preferred
ThermalSeal Sealing Film, Sterile, Prevents vapor loss and is thermostable and functional from -40deg. to 125deg.C at 75% humidity
Catalog No. NC9333898 Supplier Andwin Scientific Supplier No. T24176
ThermalSeal Sealing Film, Sterile, Prevents vapor loss and is thermostable and functional from -40deg. to 125deg.C at 75% humidity