Learn More
Alkali Scientific ThermaSeal SF2™ Sealing Film, 100/pk
SDP
Catalog No. 502389021
Encompass_Preferred
Ideal for use with flat-top, semi-skirted and full-skirted PCR plates. Certified DNase- & RNase- free. Large cut allows for easier positioning of the film to the plate via edge of plate alignment vs. centering. Tips: Selection of the proper film size will minimize evaporation. The film should not overhang or ride up the wall of a raised-rim plate. More complete coverage of skirted flat-top plates will reduce evaporation in the outer wells. End-tab removal before thermal cycling is highly recommended.
Catalog No. 50-238-9021 Supplier Alkali Scientific Supplier No. SF31ML
Ideal for use with flat-top, semi-skirted and full-skirted PCR plates. Certified DNase- & RNase- free. Large cut allows for easier positioning of the film to the plate via edge of plate alignment vs. centering. Tips: Selection of the proper film size will minimize evaporation. The film should not overhang or ride up the wall of a raised-rim plate. More complete coverage of skirted flat-top plates will reduce evaporation in the outer wells. End-tab removal before thermal cycling is highly recommended.