Learn More
Thermco Products Inc NIST Traceable Thermometer, -100/50C x 1.0C, Spirit Thermometer, 76mm Imm, 305mm long, Factory Certified @ -78, -20, 0, 4, 25, 37C
Catalog No. NC0758274
Encompass_Preferred
Thermco Certified Thermometers are used in temperature verification and calibration procedures. These instruments can be used to calibrate Liquid-In- Glass, RTD, Thermistors, Thermocouples, and Bi-Metal Thermometers. The certified thermometers are carefully inspected to insure they conform to exact specifications and are certified against NlST standards. Each thermometer is furnished with a document proving traceability and a felt-lined leatherette storage case.
Catalog No. NC0758274 Supplier Thermco Products Inc Supplier No. ACC6033FC
Thermco Certified Thermometers are used in temperature verification and calibration procedures. These instruments can be used to calibrate Liquid-In- Glass, RTD, Thermistors, Thermocouples, and Bi-Metal Thermometers. The certified thermometers are carefully inspected to insure they conform to exact specifications and are certified against NlST standards. Each thermometer is furnished with a document proving traceability and a felt-lined leatherette storage case.