Learn More
Thermco Products Inc Precision Non-Hg, Taper Joint thermometer, Spirit, -10 deg./250 deg.C, 1.0C div, 50mm imm, 350mm, 10/30 taper
Catalog No. NC0684499
Encompass_Preferred
Manufactured with a 10/30 glass standard taper joint to fit in 10/30 joint of flasks and other laboratory glassware. Accurate to N.I.S.T. tolerances. Individually serialized. Thermometer may be Safety-Coated with TFE. Filled with a safe red spirit filling for safety purposes.
Catalog No. NC0684499 Supplier Thermco Products Inc Supplier No. ACCST02250S
Manufactured with a 10/30 glass standard taper joint to fit in 10/30 joint of flasks and other laboratory glassware. Accurate to N.I.S.T. tolerances. Individually serialized. Thermometer may be Safety-Coated with TFE. Filled with a safe red spirit filling for safety purposes.