Learn More
Thermco Products Inc Special application Non-Hg, Spirit, w/safety coat, 34.5 deg./46 deg.C, 0.1C div, 76mm imm, 240mm
Catalog No. NC0583144
Encompass_Preferred
These spirit-filled glass thermometer with PTFE coating meets the EPA requirements. Coliform incubator thermometers are designed to use in incubators or baths during coliform tests. Use them to monitor equipment. Each includes an individual serial number and certificate of accuracy. The ACC713BLS can be certified and made traceable to N.I.S.T.
Catalog No. NC0583144 Supplier Thermco Products Inc Supplier No. ACC713BLSSC
These spirit-filled glass thermometer with PTFE coating meets the EPA requirements. Coliform incubator thermometers are designed to use in incubators or baths during coliform tests. Use them to monitor equipment. Each includes an individual serial number and certificate of accuracy. The ACC713BLS can be certified and made traceable to N.I.S.T.