Learn More
Thermco Products Inc NIST compliant, factory certified glass Spirit thermometer, 0 deg./300 deg.F, 2.0Fdiv, 76mm imm, certified at 32 deg.F, 100 deg.F, 200 deg.F, 300 deg.F, 305mm
Catalog No. NC0691858
Encompass_Preferred
Thermco Certified Thermometers are used in temperature verification and calibration procedures. These instruments can be used to calibrate Liquid-In- Glass, RTD, Thermistors, Thermocouples, and Bi-Metal Thermometers. The certified thermometers are carefully inspected to insure they conform to exact specifications and are certified against NlST standards. Each thermometer is furnished with a document proving traceability and a felt-lined leatherette storage case.
Catalog No. NC0691858 Supplier Thermco Products Inc Supplier No. ACC6123SFC
Thermco Certified Thermometers are used in temperature verification and calibration procedures. These instruments can be used to calibrate Liquid-In- Glass, RTD, Thermistors, Thermocouples, and Bi-Metal Thermometers. The certified thermometers are carefully inspected to insure they conform to exact specifications and are certified against NlST standards. Each thermometer is furnished with a document proving traceability and a felt-lined leatherette storage case.