Learn More
Thermco Products Inc THERMCO NIST OVEN THERM
Catalog No. NC1131886
Encompass_Preferred
AccuTemp, 95/115C x 0.01C, Hg, Mercury thermometers sealed in a PTEF jacket that contains liquid & glass if breakage occurs and are housed in unbreakable plastic bottles filled with a non-toxic Bio-Safe mediun. Economical, Precise and Reliable means of measuring critical temperatures in Ovens. Each bottle thermometer have their own unique serial number and are supplied with a NIST Traceable Certificate.
Catalog No. NC1131886 Supplier Thermco Products Inc Supplier No. ACCOV700
AccuTemp, 95/115C x 0.01C, Hg, Mercury thermometers sealed in a PTEF jacket that contains liquid & glass if breakage occurs and are housed in unbreakable plastic bottles filled with a non-toxic Bio-Safe mediun. Economical, Precise and Reliable means of measuring critical temperatures in Ovens. Each bottle thermometer have their own unique serial number and are supplied with a NIST Traceable Certificate.