Learn More
New England Biolabs, Inc. Therminator™ DNA Polymerase – 1000 units
SDP
Supplier:  New England Biolabs, Inc. M0261L
Encompass
Therminator DNA Polymerase is a 9N DNA Polymerase variant with an enhanced ability to incorporate modified substrates such as dideoxynucleotides, ribonucleotides and acyclonucleotides.
Catalog No. 50-811-684